Elementary Water Cycle Poster
Elementary Water Cycle Poster, 36", 24"
Catalog # 470008-510
Supplier: KELLER BROS. & MILLER, INC.
Supplier #: 7428100
Jump to:
Product Details & Documents
Picture the water cycle.
- Clear demonstration
- Colorful representation
- Large text and pictures
Clearly demonstrate the water cycle with colorful representations of familiar objects. Bright red arrows show the interconnected nature of a stream, a rain cloud, a pond, cloud, land, and the sun. The large size of text and pictures allow easy viewing for the whole class. Developed by Rich Cymerman. Size: 24" x 36".
Specifications
- Width:36"
- Description:Elementary Water Cycle Poster
- Height:24"