Electrostatic Pompon Set
Electrostatic Pompon Set
Catalog # 470110-370
Supplier: VWR International
Jump to:
Product Details & Documents
Electrostatic Pompon Set
Two electrostatic pompons used for electrostatic experiments. A metal rod mounted on an insulated plastic stand carries a round metal plate at its top and a fringe of 40 colored nylon cords. When the plate is touched with an electrostatically charged rod, the cords become charged and spread out into a wide cone by mutual repulsion. The pompon is well insulated and will hold its charge for up to 30 minutes in dry air. This set contains a pair of pompons.
Set of two.
Each pompon is 18.5 cm high with 10.5 cm long fringes.
Base is 7 cm in diameter.
Especificaciones
- Description:Electrostatic Pompon Set