Electrostatic Materials Class Set
Electrostatic Materials Class Set 5th Ed. PSSC
Catalog # 470148-944
Supplier: VWR International
Jump to:
Product Details & Documents
Enough materials for a complete class to perform all of the electrostatics experiments included in this set.
Contents: 8 vinyl strips, 8 acetate strips, 8 polyethylene strips, 8 aluminum tubes, 8 wool cloth squares, 8 cotton squares, 8 wooden mirror supports, and one package containing graphite coated pith balls, nylon monofilament, and aluminum foil.
Specifications
- Description:Electrostatic Materials Class Set 5th Ed. PSSC