Orders LinkContactUsLinkComponent
Electronic Digital Micrometer
Electronic Digital Micrometer
0-1", 0.0001", 0.00005"
Catalog # 470110-398
Supplier:   CHICAGO BRAND CP
Número de proveedor #:  50059
Electronic Digital Micrometer
Catalog # 470110-398
Supplier:  CHICAGO BRAND CP
Electronic Digital Micrometer
Electronic Digital Micrometer
0-1", 0.0001", 0.00005"
Price
$3,691.61
Log in Para disponibilidad
Each (1UN)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Digital Display Reads Outside Dimensions to 50 Millionths
  • Auto On/Off Function
  • Memory Hold Function
  • Absolute/Relative Modes Functions
  • Tolerance Limits/Alarms and Origin Preset
  • Carbide tipped measurement surfaces
  • Includes Calibration Wrench
Instructional graphic included to prevent over tightening.
Especificaciones
  • Resolution:
    0.00005"
  • Measurement Range:
    0-1"
  • Accuracy:
    0.0001"
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 