Orders LinkContactUsLinkComponent
Electromagnet Set
Electromagnet Set
Kit, Electromagnet Set
Catalog # 470004-120
Supplier:   DOWLING MAGNETS,LLC
Número de proveedor #:  731102
Electromagnet Set
Catalog # 470004-120
Electromagnet Set
Electromagnet Set
Kit, Electromagnet Set
Price
$1,066.02
Log in Para disponibilidad
Kit de 1
Product Details & Documents
Your students can use the Electromagnet Science Set to build an electromagnet, a doorbell, a chime, a telegraph, and more.
  • Build electromagnetic items
  • Contains a variety of reusable parts
  • Manual included
Kit contains a variety of basic, reusable parts, including a compass, an assortment of magnets and magnetic wire, switch plates and sender plates, colored lights, pre-drilled wood blocks, and a light-emitting diode, that can be used to complete several introductory experiments and six projects in electromagnetism.
A manual and a list of additional items that may be needed are also included.
Caution: WARNING: This product contains a small magnet(s). Swallowed magnets can stick together across intestines causing serious infections and death. Seek immediate medical attention if magnet(s) are swallowed or inhaled.
Especificaciones
  • Description:
    Kit, Electromagnet Set