Electric Plume
Electric Plume, Accessory for Electrostatic Generator (Van de Graaff)
Catalog # 470003-972
Supplier: SCIENCE INTERACTIVE GROUP
Supplier #: N-125
Jump to:
Product Details & Documents
Graphically illustrate the fact that like charges repel each other.
The multi-colored ribbons are connected to a suction cup that sits on top of a Van de Graaff generator. As the generator operates, the ribbons accumulate a charge and stand on end.
Specifications
- Description:Electric Plume, Accessory for Electrostatic Generator (Van de Graaff)