Electric Heaters
Electric Heaters, Power, watts: 200, Total Length: 6", Heated Length: 3"
Catalog # 470122-464
Supplier: GLO-QUARTZ ELECT HEATER CO INC. SE
Número de proveedor #: LHP-200
Jump to:
Product Details & Documents
Excellent for heating all acid solutions except hydrofluoric.
- Resistance element is completely sealed in a quartz tube
- Upper portion remains cool and can be held in the bare hand
- Diameter: ⁵/₁₆"
Permanently attached 6' 2-wire line cord/plug for 115 VAC or DC.
Especificaciones
- Description:Electric Heaters, Power, watts: 200, Total Length: 6", Heated Length: 3"