Elastic Cartilage, Rabbit Slide
Elastic Cartilage, Rabbit, Section, Verhoeff and Eosin
Catalog # 470182-644
Supplier: Avantor
Jump to:
Product Details & Documents
Examine an important structural tissue of the body
- Identifying features clearly distinguishable
- Rigorous quality control standards
Rabbit external ear. Stained to show elastic fibers between lacunae.
Especificaciones
- Description:Elastic Cartilage, Rabbit
- Stain:Verhoeff and Eosin
- Orientation:Section