Economy Spring Scales
470020-032
- Referencia:470020-032
- Capacidad de pesaje:5000 g (50 N), with 100 g (1.0 N) graduations
- Nº de proveedor:SB5000
Inexpensive yet accurate.
- Clear plastic case
- Gram and Newton measurements
- Pull type scale
These economical spring scales feature a clear plastic case so students can observe the mechanism. Calibrated in Grams in Newtons for the most versatile lab applications.
: Scales include an aluminum front panel that can be moved up or down for zero adjustment, metal pointer, suspension ring, and load hook.