Orders LinkContactUsLinkComponent
Economy Cardboard Insect Boxes
Economy Cardboard Insect Boxes
Supplier:   CHARLES W. KANTY
Economy Cardboard Insect Boxes
Supplier:  CHARLES W. KANTY
Certificates
Economy Cardboard Insect Boxes
Economy Cardboard Insect Boxes
1 Options Available Below
Product Details & Documents
Excellent for temporary storage or display.
  • Avalable in white only
  • Styrofoam bottom that can be used for pinning
These boxes feature a marbled green paper exterior with white glazed interior and pinning bottom. Available with solid or clear plastic cover. Size: 9³/₄W×2³/₄D×11³/₄L".
Product Family Options
Información del producto
  • Availability
    Price
  • 470022-330
    Economy Cardboard Insect Boxes, Solid Cover
    Economy Cardboard Insect Boxes, Solid Cover
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $949.30
    $949.30
    Each (1UN)
       $949.30
    Supplier Number: 301100
  • 470022-332
    Economy Cardboard Insect Boxes, Clear Cover
    Economy Cardboard Insect Boxes, Clear Cover
    Discontinued
    Supplier Number: -