Orders LinkContactUsLinkComponent
Economy Bench Pulleys
Economy Bench Pulleys
Economy Bench Pulleys, Vertical
Catalog # 470110-776
Supplier #:  PULBN2
Economy Bench Pulleys
Catalog # 470110-776
Economy Bench Pulleys
Economy Bench Pulleys
Economy Bench Pulleys, Vertical
Price
$276.10
Log in for availability
Each (1UN)
Product Details & Documents
Best Seller!
  • Good quality
Both pulleys feature a built-in table clamp, a 50mm (2") diameter plastic pulley, and a sturdy steel frame. The vertical model has a 30mm (1 3/16") clamp opening, and the horizontal model has a 25mm (1") opening.
Specifications
  • Description:
    Economy Bench Pulleys, Vertical