Economy Bench Pulleys
Economy Bench Pulleys, Vertical
Catalog # 470110-776
Supplier: UNITED SCIENTIFIC SUPPLIES
Supplier #: PULBN2
Jump to:
Product Details & Documents
Best Seller!
- Good quality
Both pulleys feature a built-in table clamp, a 50mm (2") diameter plastic pulley, and a sturdy steel frame. The vertical model has a 30mm (1 3/16") clamp opening, and the horizontal model has a 25mm (1") opening.
Specifications
- Description:Economy Bench Pulleys, Vertical