Economy Alcohol Lamp Stand
Catalog # 470019-486
Supplier: UNITED SCIENTIFIC SUPPLIES
Specifications
- Descripción:Economy alcohol lamp stand
- Referencia:470019-486
- Nº de proveedor:ABSP05
Specifications
About this item
This rugged wire support stand has a heavy wire gauze top.
- Wire gauze surface measures 8×10 cm (3×2")
- Height: 11.5 cm (4½")
Made especially for alcohol burners.