Orders LinkContactUsLinkComponent
Easy Grip Fine-Point Forceps
Easy Grip Fine-Point Forceps
Easy Grip Fine-Point Forceps
Easy Grip Fine-Point Forceps
Easy Grip Fine-Point Forceps
Minimize hand fatigue.
  • Large gripping area
  • Fine and medium points available
  • Straight or curved tip selection
Comfortable to handle and incredibly easy to maneuver, these stainless steel forceps feature finger grips designed to prevent slipping and provide better control when working with specimens. Available in fine point, or medium point styles.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
  • Description
    Material
    Length
    Tip Style
  • Easy Grip Fine-Point Forceps
    Stainless steel
    101 mm (4")
    Straight
    Each (1UN)
    Each (1UN) - $97.01
    $97.01
    Caja de 50 (1UN)
    Caja de 50 (1UN) - $105.74
    $105.74
    Each (1UN)
       $97.01
    Caja de 50 (1UN)
       $105.74
     : 140590
  • Easy Grip Fine-Point Forceps
    Stainless steel
    101 mm (4")
    Curved
    Each (1UN)
    Each (1UN) - $97.01
    $97.01
    Each (1UN)
       $97.01
     : 140591