Earthworm Bedding
Earthworm Bedding, 680 g 1.5lbs (24oz)
Catalog # 470006-308
Supplier: MAGIC PRODUCTS INC SE
Número de proveedor #: 100
Jump to:
Product Details & Documents
Consisting of sphagnum peat moss and organic fibers, this clean, odorless, long-lasting bedding won’t pack down.
- Organic fibers
- Odorless
- Holds up to 20X its weight
Because it holds up to 20 times its own weight in moisture, it provides an ideal environment for worms.
Especificaciones
- Description:Earthworm Bedding, 680 g 1.5lbs (24oz)