Orders LinkContactUsLinkComponent
Dual Magnifier
Dual Magnifier
Dual magnifier
Catalog # 470016-340
Número de proveedor #:  6237001
Dual Magnifier
Catalog # 470016-340
Dual Magnifier
Dual Magnifier
Dual magnifier
Price
$13.32
Log in Para disponibilidad
Each (1UN) Caja de 360 (1UN)
Product Details & Documents
The perfect tool for investigation
  • Available as a single or a pack of 10
Use this dual 3X/6X plastic magnifier to get a closer look at the world around you. Economically priced, this plastic magnifier is virtually unbreakable, and the dual magnificataion makes this a perfect tool for any investigation. The handheld magnifier features a 3X lens that is 4.5cm in diameter and a 6x lens that is 1.7cm in diameter. The overall length is 11.5cm.
Especificaciones
  • Magnification:
    3×, 6×
  • Description:
    Dual magnifier