Dressing Stick for Cutoff Saws
Dressing Stick for Cutoff Saws, 1×1×6"
Catalog # 470022-254
Supplier: SAINT GOBAIN ABRASIVES
Número de proveedor #: 281927
Jump to:
Product Details & Documents
Keeps diamond blades free of buildup.
Blades last longer and cut faster with this specially bonded silicon carbide.
Especificaciones
- Description:Dressing Stick for Cutoff Saws
- Dimensions:1×1×6"