Double-Safe™ Thermometers
Double-Safe™ Thermometers, Range: -20 to +260°C, Length, mm: 305, Immersion: Partial, Each
Catalog # 470123-612
Supplier: BEL-ART PRODUCTS SE
Número de proveedor #: B60400-0800
Jump to:
Product Details & Documents
Glass thermometers with Enviro-Safe Black liquid.
Extra-wide bore; Yellow background; Teflon® coated. 1°C Subdivisions.
Especificaciones
- Description:Double-Safe™ Thermometers, Range: -20 to +260°C, Length, mm: 305, Immersion: Partial, Each