ContactUsLinkComponent
Double Buret Clamp with Vinylized Jaws
  470121-774
undefined
Double Buret Clamp with Vinylized Jaws
  470121-774
  • Descripción:
    Double Buret Clamp with Vinylized Jaws
  • Referencia:
    470121-774
  • Nº de proveedor:
    CH0258B
Grasps any buret firmly from micro to 100 ml size.
Each buret is held independently. Locks to support rod by thumbscrew, which is tightened from the front; will fit support rods up to 13mm (1/2") in diameter.
Vinylized jaws provide nonslip grip and resist attack by acids, alkalies or solvents.