Orders LinkContactUsLinkComponent
Double Buret Clamp, Polyethylene
Double Buret Clamp
  470148-978
Double Buret Clamp, Polyethylene
  470148-978
Double Buret Clamp, Polyethylene
Double Buret Clamp, Polyethylene
Double Buret Clamp
$97.82
Each (1UN) Case de 12 (1UN)
This clamp is compatible with any 1/2" or 3/8" standard support rod and with burets from microsize to 100 ml.
Its concealed spring steel holds burets without obscuring the graduations or the meniscus.
  • Description:
    Double Buret Clamp