Donut Magnet
Magnet, Donut 3 cm OD, 1 cm ID (grade 5 strength)
Catalog # 470188-554
Supplier: MASTER MAGNETICS INC SE
Supplier #: CR145C
Jump to:
Product Details & Documents
Good for magnetic experiments. Not so good with coffee.
Specifications
- Description:Magnet, Donut 3 cm OD, 1 cm ID (grade 5 strength)
- Cat. No.:470188-554