Disposable Thermometers
Disposable Thermometers, +96,0...+104,8 °F
Catalog # 470145-068
Supplier: MEDICAL INDICATORS,INC
Supplier #: 1112-20
Jump to:
Product Details & Documents
These affordable, single-use thermometers are accurate yet eliminate the need for sterilization between uses.
- For Oral or Axillary Readings
- Individually Wrapped
- Range: 96 to 104.8°F
The last dot on the scale to turn from green to black indicates the correct temperature, and the results work for either oral or axillary readings.
The mercury-free, individually wrapped thermometers have a range of 96.0 to 104.8°F in 0.2° increments.
Specifications
- Description:Disposable Thermometers
- Range:+96,0...+104,8 °F