Disposable Micropipet Tips
Supplier: FISCHER TECHNICAL CO SE
Jump to:
About this item
High quality micropipette tips to meet any lab's needs.
- Certified DNase, RNase and pyrogen free
- Material: PP
- Autoclavable
- Designed to fit most standard micropipettes
All quantities are supplied in bulk bags.
5 Options Available Below
Product Details & Documents
High quality micropipette tips to meet any lab's needs.
- Certified DNase, RNase and pyrogen free
- Material: PP
- Autoclavable
- Designed to fit most standard micropipettes
All quantities are supplied in bulk bags.
Product Family Options
Bulk Bags
Product Information
- VolumeTip ColorPackedAvailabilityPrice