Orders LinkContactUsLinkComponent
Disposable Impression Trays
Disposable Impression Trays
Disposable Impression Trays
Certificates
Disposable Impression Trays
Disposable Impression Trays
2 Options Available Below
Product Details & Documents
Inexpensive Recreations for Labwork
  • Autoclavable
  • Easy to Clean
Cast impressions of teeth to identify placement and study bite marks for forensic odontology lessons. This set of disposable dental impression trays is autoclavable and available for upper and lower jaw impressions. Set
of 12.
Product Family Options
Información del producto
  • Includes
    Availability
    Price
  • 470020-458
    Disposable Impression Trays, Upper
    Disposable Impression Trays, Upper
    12 Trays
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 12
    Paquete de 12 - $279.82
    $279.82
    Paquete de 12
       $279.82
    Supplier Number: 1003
  • 470020-462
    Disposable Impression Trays, Lower
    Disposable Impression Trays, Lower
    12 Trays
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 12
    Paquete de 12 - $279.82
    $279.82
    Paquete de 12
       $279.82
    Supplier Number: 1004