Polyethylene Dispensing Jug
Polyethylene Dispensing Jug, 5 Gallon Capacity
Catalog # 470153-632
Supplier: BEL-ART PRODUCTS SE
Número de proveedor #: H11850-0000_SE
Jump to:
Product Details & Documents
Conveniently store up to 5 gallons of liquid that can be dispensed directly from the jug.
- Size: 9⁷/₈L×9⁷/₈W×15" H
A spigot overhangs your work surface for easy access. It is constructed of high-density polyethylene with rounded corners and a sturdy handle and bottom to easily pour large quantities.
Especificaciones
- Description:Polyethylene Dispensing Jug, 5 Gallon Capacity