Orders LinkContactUsLinkComponent
Dispensing Bottles with Hinged Caps
Dispensing Bottles with Hinged Caps
Supplier:   DYNALON
Dispensing Bottles with Hinged Caps
Supplier:  DYNALON
Certificates
Dispensing Bottles with Hinged Caps
Dispensing Bottles with Hinged Caps
2 Options Available Below
Product Details & Documents
Virtually Unbreakable.
  • Sturdy polyethylene construction
  • Hinged dropper closures included
Simply squeeze to dispense indicator solutions, dyes, etc. The hinged tops can't get lost.
Product Family Options
Información del producto
  • Capacity
    Availability
    Price
  • 30 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $91.36
    $91.36
    Log in Para disponibilidad
    Case de 10 (1UN)
    Case de 10 (1UN) - $99.59
    $99.59
    Each (1UN)
       $91.36
    Case de 10 (1UN)
       $99.59
    Supplier Number: 524165-0030
  • 60 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $71.48
    $71.48
    Log in Para disponibilidad
    Paquete de 12 (1UN)
    Paquete de 12 (1UN) - $77.92
    $77.92
    Log in Para disponibilidad
    Case de 144 (1UN)
    Case de 144 (1UN) - $10,293.44
    $10,293.44
    Each (1UN)
       $71.48
    Paquete de 12 (1UN)
       $77.92
    Case de 144 (1UN)
       $10,293.44
    Supplier Number: 211194
  • 150 ml
    Discontinued
    Supplier Number: -
  • 250 ml
    Discontinued
    Supplier Number: 524165-0250
  • 500 ml
    Discontinued
    Supplier Number: -