Orders LinkContactUsLinkComponent
Boreal Science Digital Compound Microscopes
Boreal Science Digital Compound Microscopes
Boreal Science Digital Compound Microscopes
Certificates
Boreal Science Digital Compound Microscopes
3 Options Available Below
Product Details & Documents
In these digital compound instruments, a video camera is built into the traditional microscope chassis.
  • Motic image editing software included
  • Smartboard compatible
  • One-year manufacturer's warranty on camera
  • Boreal lifetime warranty on microscope chassis
  • VAC: 100 - 120
These rugged digital microscopes offer high resolutions. One power supply operates both the microscope and camera. The included, user-friendly Motic image editing software lets you capture images in JPG, TIFF, MIG, and BMP formats and movies in AVI format.
Digital and Digital2000 microscopes connect to a computer via a USB port. Each microscope comes with all-connecting cables.
Ordering information: This product includes a manufacturer's warranty.
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 470145-726
  • 470145-722
  • 470222-452
  • Model:
  • 470145-726
  • 470145-722
  • 470222-452
  • Stage:
  • 110×120 mm (4⅜×4¾") stage, with locked-on spring mounted stage clips, drilled and tapped to accept optional mechanical stage
  • 120×120 mm (4⅜×4⅜"), Built-in mechanical stage with low-position coaxial x-y controls
  • Large 140×135 mm (5½x5-5/16") rackless mechanical stage
  • Electrical:
  • 110 V
  • 100 - 120 V
  • 100 - 120 V
  • W×D×H:
  • 5×6½×14½"
  • 5×6½×14½"
  • 7¼×9¼×16½"
Especificaciones
Product Family Options
Información del producto
  • Magnification
    Illumination
    Availability
    Price
  • 470145-726
    Boreal monocular digital 5 MP microscope
    Boreal monocular digital 5 MP microscope
    4×, 10×, 40×, 195B
    LED cordless
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $13,863.51
    $13,863.51
    Each (1UN)
       $13,863.51
    Supplier Number: 5584700
    Especificaciones:
    More
    Specifications
    Cat. No.
    470145-726
    Model
    470145-726
    Stage
    110×120 mm (4⅜×4¾") stage, with locked-on spring mounted stage clips, drilled and tapped to accept optional mechanical stage
    Electrical
    110 V
    W×D×H
    5×6½×14½"
  • 470145-722
    Boreal monocular digital 5 MP microscope
    Boreal monocular digital 5 MP microscope
    4×, 10×, 40×, 195B
    Halogen
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $689.45
    $689.45
    Liquidación
    Each (1UN)
       $689.45
    Supplier Number: 1100400200075
    Especificaciones:
    More
    Specifications
    Cat. No.
    470145-722
    Model
    470145-722
    Stage
    120×120 mm (4⅜×4⅜"), Built-in mechanical stage with low-position coaxial x-y controls
    Electrical
    100 - 120 V
    W×D×H
    5×6½×14½"
  • 470222-452
    Boreal trinocular digital 5 MP microscope, with 30° inclined forward-facing trinocular head
    Boreal trinocular digital 5 MP microscope, with 30° inclined forward-facing trinocular head
    4×, 10×, 40×R, 100×R
    LED
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,584.90
    $1,584.90
    Liquidación
    Each (1UN)
       $1,584.90
    Supplier Number: DC5-169-ASC
    Especificaciones:
    More
    Specifications
    Cat. No.
    470222-452
    Model
    470222-452
    Stage
    Large 140×135 mm (5½x5-5/16") rackless mechanical stage
    Electrical
    100 - 120 V
    W×D×H
    7¼×9¼×16½"
Accessories
Información del producto
  • Catalog #
    Electrical
    Availability
    Price
  • Replacement bulb
    Replacement bulb
    470005-050
    1 W
    Log in Para disponibilidad
    Each (1UN) - $159.92
Información del producto
  • Catalog #
    Availability
    Price
  • Replacement Bulb 20W 12V Halogen
    Replacement Bulb 20W 12V Halogen
    470021-964
    Log in Para disponibilidad
    Each (1UN) - $391.22