Digestive System Composite Slide
Digestive System Composite Slide, Section, Hematoxylin and Eosin
Catalog # 470182-864
Supplier: Avantor
Jump to:
Product Details & Documents
Compare and contrast mammalian digestive tissue
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Three regions: stomach, or fundic portion, small intestine, and large intestine.
Especificaciones
- Description:Digestive System Composite Slide
- Stain:Hematoxylin and Eosin
- Orientation:Section