Differential and Sympathetic Tuning Forks
Catalog # 470148-808
Supplier: VWR International
Specifications
- Descripción:Differential and Sympathetic Tuning Forks
- Referencia:470148-808
- La x An x Al:30 x 22 x 27 cm
- Peso:1.3 kg (3 lbs.)
- Nº de proveedor:P36140
Specifications
About this item
These large alloy tuning forks are designed to demonstrate resonance, beat frequency, and sympathetic vibration.
- Frequency: 256Hz
- Moveable Mass Changes Frequency
- Forks Demonstrate Resonance, Beat Frequency, and Sympathetic Vibration
Each fork has a frequency of 256Hz. One fork has a movable mass to change the frequency. The closer the frequency is to the other tuning fork, the slower the beat pattern.
Students can see the beat patters using a probeware microphone. Assembly is required.