Kemtec® Dichotomous Key Kit
470102-506
- Descripción:Dichotomous Key Kit
- Referencia:470102-506
- Nº de proveedor:1-178
Learn to identify species.
- Learn to use keys for identification
- Learn to build a dichotomos key
- Reusable
Dichotomous keys are used to classify and identify objects or living organisms. Guide students through the process of using keys with several real-world examples, just like scientists. Includes seven different activities, ranging from easy to challenging and basic to scientific, allowing students of differing abilities to readily learn to use a key. Culminating activities include constructing a tree key to your own school and using a dichotomous key to identify seashells using scientific names.
: See datasheet for contents information.