Diatom Strew Slide
Diatom Strew, Explano-Mount™, Unstained
Catalog # 470177-018
Supplier: Avantor
Jump to:
Product Details & Documents
Mixed forms of phytoplankton
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mixed species. Includes pennate and centric types.
Especificaciones
- Description:Diatom Strew
- Stain:Unstained
- Orientation:Explano-Mount™