Dialysis Bag Holder
Catalog # 470201-746
Supplier: BEL-ART PRODUCTS SE
Specifications
- Descripción:Dialysis Bag Holder
- Referencia:470201-746
- Nº de proveedor:182370000
Specifications
About this item
Easy to attach.
- The autoclavable clip attaches to the side of a beaker
- Size 2" L x 0.625" W
- Package of six