ContactUsLinkComponent
Detection of Mad Cow Disease Lab Activity
Detection of Mad Cow Disease Lab Activity
Catálogo # 470007-124
Proveedor:  EDVOTEK, INC.
undefined
Detection of Mad Cow Disease Lab Activity
Catálogo # 470007-124
Proveedor:  EDVOTEK, INC.
Número de proveedor:  117
Especificaciones
  • Descripción:
    Detection of Mad Cow Disease Lab Activity
  • No. of Lab Groups:
    8
  • Referencia:
    470007-124
  • Nº de proveedor:
    117
Especificaciones
Acerca de este artículo
Examine Simulated PCR Products to Find BSE
  • Convenient Ready-To-Load samples
  • Connection to infectious disease
  • Inquiry-based experiment
  • Time required: ~45 minutes
Bovine spongiform encephalopathy (BSE), better known as mad cow disease, is a neurodegenerative, fatal condition in cattle. Consuming BSE-infected beef is believed to be the cause of a similar condition in humans, Creutzfeldt-Jakob disease. In this experiment, students examine simulated PCR products from several feed mills, to determine any possible violations of a 1997 ban which ended the practice of including animal parts in cattle feed.
Kit includes: instructions, Ready-to-Load™ QuickStrip™ DNA samples, UltraSpec-Agarose™ powder, practice gel loading solution, electrophoresis buffer, InstaStain® Blue and FlashBlue™ stain, calibrated pipette, and microtipped transfer pipettes.
Required, but not included: electrophoresis apparatus, power supply, automatic micropipette and tips, balance, microwave or hot plate, visualization system.
Embalaje: Store Ready-to-Load QuickStrip™ samples in the refrigerator