Density Tube
Density Tube
470152-828
: Avantor
Teacher Developed!
- Grades 7–12
- Size: 24.8cm H
Physics teacher Victor Noto has developed a great way to demonstrate heat transfer by convection. His density tube uses water at different temperatures to produce bulk liquid flow. By timing how long it takes for a small volume of liquid to move up to the top of the tube, students can determine if the rate of heat transfer depends on the density differences. Includes instructions.
- Description:Density Tube