Orders LinkContactUsLinkComponent
Demonstration Meter
Supplier:   VWR International
Demonstration Meter
Supplier:  VWR International
Certificates
Demonstration Meter
Demonstration Meter
3 Options Available Below
Product Details & Documents
This working meter can be used to demonstrate the working principle of AC or DC ammeters and voltmeters with the interchangeable plastic scales, available separately.
Basic sensitivity is 5mA, 100mV. It is housed in a ABS plastic case with glass front and rear. Size: 30 x 15 x 30 cm.
Scale inserts: 0-100 mA (DC); 0-300 V(DC); 0-1A (AC); 0-300 (AC). Package of four.
Scale inserts: 0-1A (DC); -10 to +10 V (DC). Package of two.
Product Family Options
Información del producto
  • Availability
    Price
  • 470039-432
    Demonstration Meter
    Demonstration Meter
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $50.05
    $50.05
    Liquidación
    Each (1UN)
       $50.05
  • 470039-434
    Demonstration Meter, Scale Inserts, Set of 4: 0-100 mA (DC); 0-300 V(DC); 0-1A (AC); 0-300 (AC);
    Demonstration Meter, Scale Inserts, Set of 4: 0-100 mA (DC); 0-300 V(DC); 0-1A (AC); 0-300 (AC);
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $863.45
    $863.45
    Liquidación
    Each (1UN)
       $863.45
  • 470039-436
    Demonstration Meter, Scale Inserts, Set of 2: 0-1A (DC); -10 to +10 V (DC)
    Demonstration Meter, Scale Inserts, Set of 2: 0-1A (DC); -10 to +10 V (DC)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $13.75
    $13.75
    Liquidación
    Each (1UN)
       $13.75