Ward's® Deluxe Model Stream Table Kits
Standard Stream Table, 20", 60", 6"
470029-374
: 105-RT
Stream table kits For the most detailed and thorough demonstrations of sediment transport.
- Hands-on learning of coastal environments
- Investigate water movement and how coastlines can change
Demonstrate coastline formation with a wavemaker, follow the course of a stream and more with this table-size stream table kit. Heavy, seamless plastic construction provides years of leakproof use.
: Replacement or additional power supplies and pump available for purchase.
: Includes a power pack and pump for variable water flow, sediment trap, internal water reservoir, and wave-maker. 125 VAC with 71/2' cord on power pack, 8' cord on pump. The combinations packs also include the rainfall simulator, terrain feature model, contour map grid, and a 32 page stream table investigation model.
- Description:Standard Stream Table
- Height:6"
- Length:60"
- Width:20"