Deluxe Metric Micrometer
0-25 mm, 0.01 mm
Catalog # 470110-220
Supplier: MTI CORP CP
Número de proveedor #: 103-137
Jump to:
Product Details & Documents
Outside Mechanical Micrometer
- Coated in smooth satin chrome
- Clearly graduated
- Includes a ratchet stop
Especificaciones
- Resolution:0.01 mm
- Measurement Range:0-25 mm