Deflagration Spoon
Deflagration Spoon
Catalog # 470121-939
Supplier: HUMBOLDT MFG CO
Supplier #: H-10400
Jump to:
Product Details & Documents
This spoon is made out of durable brass.
- Handle: 40 cm
- Bowl Diameter: 16 mm
- Brass Construction
The spoon has a 40 cm handle and a 16 mm diameter bowl.
Specifications
- Description:Deflagration Spoon