Orders LinkContactUsLinkComponent
Decomposition Lab Activity
Decomposition Lab Activity
Decomposition Lab Activity, 50
Catalog # 470039-840
Número de proveedor #:  35
Decomposition Lab Activity
Catalog # 470039-840
Decomposition Lab Activity
Decomposition Lab Activity
Decomposition Lab Activity, 50
Price
$6,618.89
Log in Para disponibilidad
Each (1UN)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Guided Inquiry Study of Soil
  • Students will create compost
  • Demonstrate biodegradability
  • Connects with other environmental concepts
Explore the concepts of biodegradability and decomposition and how they apply to the study of life cycles. This series of inquiry-based experiments investigate the effect of soil type on decomposition, the different types of substances which decompose, and demonstrate how to determine the rate of decomposition. Includes materials for 50 students.
Especificaciones
  • Description:
    Decomposition Lab Activity
  • No. of Lab Groups:
    50
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 