Cyanobacteria and Eubacteria Slide
Cyanobacteria & Eubacteria, Whole mount & smear, Composite: Multiple stains
470180-840
: Avantor
Contrast Organism Structures
- Identifying features clearly distinguishable
- Rigorous quality control standards
Comparison of blue-green algae and eubacteria.
- Description:Cyanobacteria & Eubacteria
- Stain:Composite: Multiple stains
- Orientation:Whole mount & smear