Orders LinkContactUsLinkComponent
Economy Dissection Instruments
Economy Dissection Instruments
Certificates
Economy Dissection Instruments
Economy Dissection Instruments
12 Options Available Below
Product Details & Documents
Priced for even the tightest budget.
  • Plated steel
  • Available in multiple sizes and styles
  • Great for student dissection sets
 : These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
Información del producto
  • Material
    Length
    Tip Style
    Availability
    Price
  • 470104-190
    Curved Medium Point Forceps
    Curved Medium Point Forceps
    Nickel Plated
    115 mm (4.5")
    Curved
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $35.19
    $35.19
    Each (1UN)
       $35.19
    Supplier Number: 6149000
  • 470018-870
    Dissecting Forceps
    Dissecting Forceps
    Nickel Plated
    110 mm (4.5")
    Straight
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $26.12
    $26.12
    Log in Para disponibilidad
    Caja de 250 (1UN)
    Caja de 250 (1UN) - $28.47
    $28.47
    Each (1UN)
       $26.12
    Caja de 250 (1UN)
       $28.47
    Supplier Number: 140512
  • 470121-996
    Dressing Forceps
    Dressing Forceps
    Chrome plated
    140 mm (5.5")
    Straight
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $42.12
    $42.12
    Each (1UN)
       $42.12
    Supplier Number: S26525-B
  • 470003-330
    Student Forceps
    Student Forceps
    Nickel Plated
    115 mm (4.5")
    Straight
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $28.25
    $28.25
    Each (1UN)
       $28.25
    Supplier Number: 2004800
  • 470106-332
    Thumb Forceps
    Thumb Forceps
    Nickel Plated
    115 mm (4.5")
    Straight
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $44.23
    $44.23
    Each (1UN)
       $44.23
    Supplier Number: BOREAL
  • 470005-436
    Medium Point Forceps
    Medium Point Forceps
    Nickel Plated
    115 mm (4.5")
    Straight
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $34.64
    $34.64
    Each (1UN)
       $34.64
    Supplier Number: 6148000
  • 470005-438
    Sharp Point Forceps
    Sharp Point Forceps
    Nickel Plated
    115 mm (4.5")
    Curved
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $35.19
    $35.19
    Each (1UN)
       $35.19
    Supplier Number: 6148500
  • 470104-188
    Straight Sharp Point Forceps
    Straight Sharp Point Forceps
    Nickel Plated
    115 mm (4.5")
    Straight
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $34.64
    $34.64
    Each (1UN)
       $34.64
    Supplier Number: 6147500
  • 470001-828
    Dissecting Scissors
    Dissecting Scissors
    Nickel Plated
    110 mm (4.5")
    Sharp/Sharp
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $18.66
    $18.66
    Each (1UN)
       $18.66
    Supplier Number: 140525
  • 470005-452
    Carbon Steel Scalpel
    Carbon Steel Scalpel
    Carbon steel
    152 mm (6")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $34.64
    $34.64
    Log in Para disponibilidad
    Caja de 50 (1UN)
    Caja de 50 (1UN) - $37.76
    $37.76
    Each (1UN)
       $34.64
    Caja de 50 (1UN)
       $37.76
    Supplier Number: 144320
  • 470122-000
    Student Scalpel
    Student Scalpel
    Nickel Plated
    128 mm (5")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $31.44
    $31.44
    Each (1UN)
       $31.44
    Supplier Number: WLS26640
  • 470147-464
    Student Scalpel
    Student Scalpel
    Nickel Plated
    128 mm (5")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $51.16
    $51.16
    Each (1UN)
       $51.16
    Supplier Number: 24