Curved Forceps
Curved Forceps
Catalog # 25729-810
Supplier: GROBET FILE CO OF AMERICA INC
Número de proveedor #: 57.907
Jump to:
Product Details & Documents
Stainless steel forceps are useful for handling small weights. Length: 10cm.
Especificaciones
- Description:Curved Forceps