Orders LinkContactUsLinkComponent
Culture Flasks
Culture Flasks
Culture Flasks
Supplier:  CORNING CA
Certificates
Culture Flasks
1 Options Available Below
Product Details & Documents
Specially Coated Tissue Culture Flasks.
  • Nunclon surface treatment promotes cell attachment and growth
  • Easy filling and access for pipets
  • Stackable, space saving design
Cell adherence is discouraged from the untreated sidewalls, neck and top panel.
Product Family Options
Información del producto
  • Availability
    Price
  • 50 mL
    Discontinued
    Supplier Number: 163371
  • 600 mL
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 5
    Paquete de 5 - $708.00
    $708.00
    Log in Para disponibilidad
    Case de 10 (5UN)
    Case de 10 (5UN) - $1,305.79
    $1,305.79
    Paquete de 5
       $708.00
    Case de 10 (5UN)
       $1,305.79
    Supplier Number: 430823