Cucurbita Stem Combination Slide
Cucurbita Stem Combination, Cross section & longitudinal section, Quadruple stained
Catalog # 470177-156
Supplier: Avantor
Jump to:
Product Details & Documents
Illustrates Structure of Gourd Plant Stem
- Identifying features clearly distinguishable
- Rigorous quality control standards
Shows both sieve plates and tubes.
Especificaciones
- Description:Cucurbita Stem Combination
- Stain:Quadruple stained
- Orientation:Cross section & longitudinal section