Square Glass Coverslips
Supplier: UNITED SCIENTIFIC SUPPLIES
Jump to:
About this item
Coverslip One is 0.15 mm thick. There are approximately 100–115 coverslips per ounce.
- Non corrosive glass
- Pre-cleaned
Coverslip Two is 0.2 mm thick. There are approximately 75–100 coverslips per ounce.
3 Options Available Below
Product Details & Documents
Coverslip One is 0.15 mm thick. There are approximately 100–115 coverslips per ounce.
- Non corrosive glass
- Pre-cleaned
Coverslip Two is 0.2 mm thick. There are approximately 75–100 coverslips per ounce.
Certificates
Search by Lot NumberProduct Family Options
Product Information
- L×WThicknessNumberingAvailabilityPrice
- 470019-002Glass cover slips, squareSupplier Number: SCSS18-1OZ
- 470019-004Glass cover slips, squareSupplier Number: SCSS22-1OZ
- 470145-754Glass cover slips, squareSupplier Number: 94-25252
- 470151-072Glass cover slips, squareGlass cover slips, square18×18 mm0.20 mm#2Discontinued-Supplier Number: -
- Discontinued-Supplier Number: -
- Discontinued-Supplier Number: -