Orders LinkContactUsLinkComponent
Ward's® Corundum
 :   Avantor
Ward's® Corundum
 :  Avantor
Ward's® Corundum
Ward's® Corundum
Crystals and crystal sections up to 1", Mohs’ hardness of 9.
  • Collected in India
  • Multiple sizes and pack options to fit your specific needs
  • Prepped by Ward's geologists to guarantee top market quality
Student Specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Bulk Pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.5×1.5 to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Product Family Options
Specimens
  • Description
    Specimen Dimensions
  • Corundum (crystals), student specimens
    <1"
    Paquete de 10
    Paquete de 10 - $668.40
    $668.40
    Paquete de 10
       $668.40
  • Corundum (crystals), bulk pack, 1 kg
    Size will vary
    Each (1UN)
    Each (1UN) - $3,787.80
    $3,787.80
    Each (1UN)
       $3,787.80