Cork Bore Sets with Handles
470202-868
- Ø orificio:5 - 24 mm (³⁄₁₆ - ¹⁵⁄₁₆")
- Descripción:Cork borers with handles
- Referencia:470202-868
- Nº de proveedor:H-9665
Bore your own holes in corks or rubber stoppers. Each set begins with a 5 mm size and increases in steps of approximately 1.3 mm.
The handles are numbered for easy identification, and a punch is included in each set.