Orders LinkContactUsLinkComponent
Coplin Staining Jars
Coplin Staining Jars
Supplier:   VARIETY GLASS INC
Coplin Staining Jars
Supplier:  VARIETY GLASS INC
Certificates
Coplin Staining Jars
Coplin Staining Jars
2 Options Available Below
Product Details & Documents
Coplin Jar with Screw-on Top Prevents Spills.
  • Quality slide making supplies
  • Slide staining tools
The screw-on cover prevents evaporation and contamination when staining. Each jar holds five single slides or ten back to back.
Product Family Options
Información del producto
  • Capacity
    Availability
    Price
  • 470175-194
    Coplin Staining Jar, Glass
    Coplin Staining Jar, Glass
    Up to 10 Slides
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $429.96
    $429.96
    Log in Para disponibilidad
    Case de 12 (1UN)
    Case de 12 (1UN) - $468.66
    $468.66
    Each (1UN)
       $429.96
    Case de 12 (1UN)
       $468.66
    Supplier Number: 674
  • 470174-652
    Coplin Staining Jar, Plastic
    Coplin Staining Jar, Plastic
    Up to 10 Slides
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $85.46
    $85.46
    Log in Para disponibilidad
    Case de 12 (1UN)
    Case de 12 (1UN) - $93.15
    $93.15
    Each (1UN)
       $85.46
    Case de 12 (1UN)
       $93.15
    Supplier Number: 62101-PK/12