Orders LinkContactUsLinkComponent
Constant Motion Machine
Constant Motion Machine
Constant motion machine, 18×10×6.5 cm
Catalog # 470148-714
Supplier:   VWR International
Constant Motion Machine
Catalog # 470148-714
Supplier:  VWR International
Constant Motion Machine
Constant Motion Machine
Constant motion machine, 18×10×6.5 cm
Price
$510.58
Log in for availability
Each (1UN) Case de 24 (1UN)
Product Details & Documents
Students construct simple motion experiments with this machine.
  • Demonstrate the relationship between velocity, displacement and time
  • Compact but powerful
  • 18×10×6.5 cm
  • Runs on two AA batteries
Demonstrate the principles of uniform linear motion using this powerful 'toy'. Moving with a constant speed, this vehicle demonstrates the relationship between velocity, displacement and time. The vehicle can also tow dynamics carts to demonstrate vectors. AA batteries make this a convenient alternative to other vehicles.
Ordering information: Two AA batteries required, but not included.
Specifications
  • Description:
    Constant motion machine
  • L×W×H:
    18×10×6.5 cm
Accessories
Product Information
  • Catalog #
    Type
    Voltage
    Availability
    Price
  • Alkaline battery 1.5 V
    Alkaline battery 1.5 V
    470156-626
    AA
    1.5 V
    Log in for availability
    Each (1UN) - $12.36