Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Especificaciones
- Descripción:Conductivity of Solutions Experiment
- No. of Lab Groups:15
- Referencia:470015-250
- Incluye:Teacher Guide, SDS, Student Guide, Worksheets
- Nº de proveedor:313
Especificaciones
Acerca de este artículo
This kit includes a series of fun activities that lead to a better understanding of the concept of ionization.
- Study principles of ionization
- Understand strong and weak electrolytes
- Differentiate between strong and weak acids and bases
- Requires one 50 minute class period to complete
Using LAB-AIDS® Conductivity Indicators, students study the principles of ionization by differentiating between strong electrolytes, weak electrolytes and non-electrolytes. They also learn to differentiate between strong and weak acids and strong and weak bases. Performing a series of fun activities, students observe and discover dramatic results which lead to a better understanding of the principles of ionization. This Lab-Aid is designed for use by students in introductory chemistry courses and higher.