Compound Bar
Compound Bar, 30 cm Length
Catalog # 470005-408
Supplier: AMERICAN SCIENTIFIC LLC SE
Número de proveedor #: 3230-00
Jump to:
Product Details & Documents
Laminated for long use.
- Bar length is 20 cm
- Bar and handle length is 30 cm
A laminated bar of two metals fastened in a wooden handle are placed over a flame. The different thermal expansion rates cause the laminated bar to bend.
Especificaciones
- Description:Compound Bar, 30 cm Length